Safety burners with stopcock
Lieferant: Usbeck
451-1369EA 122 EUR
451-1369 USBE1341
Safety burners with stopcock
Heizgeräte Gasbrenner
This safety burner provides protection against uncontrolled escape of gas in laboratories. If the flame is extinguished, the gas supply is interrupted and no unburnt gas can escape.
  • Safety pilot flame
  • 90° stopcock
  • With air regulation and thermocouple