Zinkbromid Hydrat ≥99,9% (Metall-Basis)

Lieferant: Thermo Scientific
Gefahr

44230.22EA 66.4 EUR
44230.22 44230.36
Zinkbromid Hydrat ≥99,9% (Metall-Basis)
Zinkbromid Hydrat
Formel: Br₂Zn·H₂O
Molecular Weight: 225,19 g/mol (anhy)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011294
CAS-Nummer: 7699-45-8
EINECS: 231-718-4
UN: 3260
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR