Wasser, Zellkultur-Qualität pyrogenfrei
Lieferant: VWR Chemicals
Synonyme:
Wasserstoffoxid
, Hydrogenoxid
392-0438EA
9.1
EUR
392-0438
392-0439
Wasser, Zellkultur-Qualität pyrogenfrei
Wasser
Cell culture water is high quality water suitable for use as a solvent in the preparation of cell culture media and laboratory reagents.
- Pyrogen-free
Suitable for dissolving dry powder cell culture media in accordance with the technical data sheet.
Bitte beachten The product is intended to be used in vitro, in laboratory only. Do not use it in therapy, human or veterinary applications.
Formel:
H₂O Molecular Weight: 18,02 g/mol Siedepunkt: 100 °C (1013 hPa) Schmelzpunkt: 0 °C Dichte: 1 g/cm³ (4 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011332 CAS-Nummer: 7732-18-5 EINECS: 231-791-2 Merck Index: 12,10175 |
Spezifikation Testergebnisse
Color | colourless, clear solution |
Endotoxins | < 0.005 EU/ml |
Sterility test | Bacteria in aerobic and anaerobic conditions |
Sterility test | Fungi and yeasts |
Shelf life | 48 months |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...