Corning® ITS Universal Culture Supplement Premix, Corning
Lieferant: Corning
Corning®
392-0320EA 463.13 EUR
392-0320 734-1315 392-2505
Corning® ITS Universal Culture Supplement Premix, Corning
Zellkultur-Ergänzungen und -Zusätze
ITS contains insulin, human transferrin, and selenous acid, the three most universally essential components of defined culture media.
They stimulate cell proliferation of a variety of cells under serum-reduced conditions.
Tested for ability to promote proliferation of HeLa cells in serum-free medium. Filtered (0.2 µm membrane) and tested and found negative for the presence of bacteria, fungi and mycoplasma.