Protein Coated Cell Culture Vessels, Poly-Lysine CELLCOAT®
Lieferant: Greiner bio-one
CELLCOAT®
392-0045EA 705 EUR
392-0045 392-0046 392-0134 392-0277 392-0285 392-0275 737-0080 392-0027 392-0038 392-0039 392-0028 392-0135 738-0014 738-0012 738-0013 392-0280 737-0078 737-0079 392-0273 392-0270 392-0282
Protein Coated Cell Culture Vessels, Poly-Lysine CELLCOAT®
Mikrotiterplatten Zellkultur-Platten Zellkulturplatten, beschichtet
PS, coated with poly-lysine. Poly-D-Lysine (PDL) and Poly-L-Lysine (PLL) are synthetic molecules that are used to improve the adhesion of different cell types to polystyrene surfaces. The cultivation efficiency of individual cell lines can be improved, especially when serum-free or serum-reduced medium is used, or when experiments such as transfections are performed. As the PDL is synthetic it is free of the impurities carried by other proteins.
  • Improved cell adhesion, proliferation and growth of transfected cell lines (eg. HEK-293, PC12, L929, certain 3T3 cell lines), neuronal cell lines, as well as primary neurons and glia cells
  • Suitable for serum-free or serum-reduced cell cultivation
  • Quality control tested to demonstrate promotion of the adhesion of RCG cells (rat neurons from the cerebellum)
Molecular weight: PDL 75 to 150 kD; PLL 30 to 70 kD.