Cell culture plates, Advanced TC™
Lieferant: Greiner bio-one
Advanced TC™
392-0242EA 566 EUR
392-0242 392-0231 392-0246 392-0249
Cell culture plates, Advanced TC™
Mikrotiterplatten Zellkultur-Platten
Microplates with a modified culture surface to optimise cell characteristics and functions. The improved adherence and increased growth rate improve and accelerate cell proliferation. Additionally, the Advanced TC™ surface facilitates uniform and consistent cell adherence, increases throughput and reduces cell loss, for example during automated washing stages. Free of detectable DNase, RNase and human DNA.
  • Increased cell adherence
  • Morphology similar to in vivo conditions
  • Optimal cultivation conditions for sensitive cells
  • Enables reduced quantities of serum or serum-free use
  • Free of cytotoxic substances