96- and 384-well cell culture microplates, Advanced TC™
Lieferant: Greiner bio-one
Advanced TC™ Small Volume™
392-0265EA 734 EUR
392-0265 392-0230 392-0264 392-0269 392-0248 392-0268 392-0229 392-0227 392-0228
96- and 384-well cell culture microplates, Advanced TC™
Mikrotiterplatten Zellkultur-Platten
Microplates with a modified culture surface to optimise cell characteristics and functions. The improved adherence and increased growth rate improve and accelerate cell proliferation. Additionally, the Advanced TC™ surface facilitates uniform and consistent cell adherence, increases throughput and reduces cell loss, for example during automated washing stages. Free of detectable DNase, RNase and human DNA.
  • Increased cell adherence
  • Morphology similar to in vivo conditions
  • Optimal cultivation conditions for sensitive cells
  • Enables reduced quantities of serum or serum-free use
  • Free of cytotoxic substances