Cell culture dishes, Advanced TC™
Lieferant: Greiner bio-one
Advanced TC™
392-0221EA 0 EUR
392-0221 392-0222 392-0220 392-0245 392-0217
Cell culture dishes, Advanced TC™
Zellkulturschalen Zell- und Gewebekultur-Platten
  • Enhanced cell attachment and higher proliferation rates improve and accelerate cell expansion
  • Surface treatment facilitates consistant and homogenous cell attachment increasing the overall cell yield and reducing cell loss, for example during automated washing steps
  • The chemical modification of the polymer coating assures consistant and reproducible product quality
For the propagation of fastidious cells like primary or sensitive cells as well as cells cultivated under restricted growth conditions (serum-free or serum-reduced).
  • Enhanced cell attachment and higher proliferation rates improve and accelerate cell expansion
  • Surface treatment facilitates consistent and homogenous cell attachment increasing the overall cell yield and reducing cell loss, for example during automated washing steps
  • The chemical modification of the polymer coating assures consistant and reproducible product quality