Cell Culture Roller Bottles, CELLMASTER®
Lieferant: Greiner bio-one
CELLMASTER®
GREI680058_24EA 220 EUR
GREI680058_24 392-0065 392-0066 GREI680068_24 392-0070 392-0082 GREI680065_24 392-0063 GREI681075_24
Cell Culture Roller Bottles, CELLMASTER®
Flaschen Rollerflaschen
PS with screw caps (HDPE) or filter screw caps with hydrophobic membrane (PET/PTFE), cell culture treated surface, sterile.
Available in short or long form and with a smooth or a ribbed surface. The ribbed design increases the growth area of the roller bottle without changing the height. Short form bottles with a ribbed design have a 2-fold or 2.5-fold expanded growth area. Long forms with a ribbed design have a 5—fold expanded growth area compared to the smooth form. All bottles are certified free of RNase, DNase and human DNA, are non-pyrogenic and non-cytotoxic.
  • Improved cell adhesion due to physical surface treatment
  • High stability and optical clarity
  • Thread enables quick opening with a ⅔ turn
  • Screw caps have larger knurls for improved grip and ease of opening/closing
  • Lot number and best-before date on each roller bottle
Available in short or long form and with a smooth or a ribbed surface. The ribbed design increases the growth area of the roller bottle without changing the height. Short form bottles with a ribbed design have a 2-fold or 2,5-fold expanded growth area. Long forms with a ribbed design have a 5—fold expanded growth area compared to the smooth form. All bottles are certified free of RNase, DNase and human DNA, are non-pyrogenic and non-cytotoxic.
  • Improved cell adhesion due to physical surface treatment
  • High stability and optical clarity
  • Thread enables quick opening with a ⅔ turn
  • Screw caps have larger knurls for improved grip and ease of opening/closing
  • Lot number and best-before date on each roller bottle
Ø×H: Short form 122×271 mm; long form 122×500 mm
Ø×H: Short form 122×271 mm; long form 122×500 mm
Standards: USP Class VI certified
Bitte beachten *Produkte auf dieser Seite sind außerhalb des Anwendungsbereichs der EU Richtlinie 2019/904