Cell culture dishes, glass bottom, CELLview™
Lieferant: Greiner bio-one
CELLview™ Advanced TC™ CELLview™
391-0256EA 469 EUR
391-0256 391-0255 391-0257 391-0252 391-0254 391-0253
Cell culture dishes, glass bottom, CELLview™
Zellkulturschalen Zell- und Gewebekultur-Platten
PS with integrated glass bottom, sterile. CELLview™ glass bottom dishes combine the convenience of a standard 35 mm disposable plastic cell culture dish with the optical quality of glass, providing superior high resolution microscopic images of in vitro cultures. The CELLview™ dish is also available as a subdivided version with four individual compartments facilitating simultaneous multiplex analysis of different cell lines, various stimulations or diverse transfections, minimising the amount of required cells and reagents per individual assay.
CELLview™ glass bottom dishes combine the convenience of a standard 35 mm disposable plastic cell culture dish with the optical quality of glass, providing superior high resolution microscopic images of in vitro cultures. The CELLview™ dish is also available as a subdivided version with four individual compartments facilitating simultaneous multiplex analysis of different cell lines, various stimulations or diverse transfections, minimising the amount of required cells and reagents per individual assay.
  • High transparent achromatic borosilicate glass base; hydrolytic class 1 (DIN-ISO 719)
  • Maximal spectral transmission; no autofluorescence; no depolarisation of light
  • Available with specific surface treatments increasing the attachment of adherent cells
  • Broad range of multiple microscopic applications
  • High transparent achromatic borosilicate glass base; hydrolytic class 1 (DIN-ISO 719)
  • Maximal spectral transmission; no autofluorescence; no depolarisation of light
  • Available with specific surface treatments increasing the attachment of adherent cells
  • Broad range of multiple microscopic applications
Manufactured according to ISO 8255-1:1986
Standards: Manufactured according to ISO 8255-1:1986