Paramat, Erstarrungspunkt: 55 - 58 °C, Pastillenform, Gurr™, ohne DMSO

Lieferant: VWR Chemicals

361144VEA 55.8 EUR
361144V 361148C 361147B
Paramat, Erstarrungspunkt: 55 - 58 °C, Pastillenform, Gurr™, ohne DMSO
Paraffinwachs versetzt mit synthetischen Polymeren

Paramat is a finely filtered blend of specially selected high purity paraffin wax with an addition of polymer to give improved embedding, tissue support and reliable sectioning. Residual moisture in the tissue and traces of intermedium are dispelled. The pastille shape makes for easy handling.


  • Paraffin wax blended with synthetic polymers
  • Solidification point 55 - 58 ° C


Available also with DMSO - see Paramat Extra (VWRC36133)

CAS-Nummer: 8002-74-2

Bestellen

Spezifikation Testergebnisse

Cloud point (65 °C) Clear
Cutting/ribboning Complies
Congealing temperature 55 - 58 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR