Blood collection needles, VACUETTE®
Lieferant: Greiner bio-one
VACUETTE®
GREI450073_2000EA 218 EUR
GREI450073_2000 GREI450066_50 GREI450066_1000 GREI450077_100 GREI450065_1000 GREI450072_100 331-4171 GREI450059_100 331-4170 331-4172 GREI450075_100 GREI450153_50
Blood collection needles, VACUETTE®
Injektionsnadeln
These needles are designed for blood collection procedures.
  • Colour-coded for simple identification of needle gauge
  • Designed exclusively for single use
  • Single-sample needles for veterinary use only