C1-C6 n-Paraffins, each component in nitrogen, analytical standards, Supelco®
Lieferant: Merck
SUPL501840EA 198 EUR
SUPL501840 SUPL501786 SUPL501859 SUPL23444 SUPL501778 SUPL330300
C1-C6 n-Paraffins, each component in nitrogen, analytical standards, Supelco®
Multi-component gas mixture suitable for GC calibration.
  • A certificate of analysis is included.
  • Packaged in lightweight, non-refillable, recyclable containers.
  • Portable, convenient, and easy to use.
  • Manufactured gravimetrically for maximum accuracy.
  • NIST traceable.