qPCR master mixes, Fast Plus EvaGreen®
Lieferant: Biotium
EvaGreen®
31020-TEA 135 EUR
31020-T 31020-2 31020-1 31015. 31014. 31014-T 31015-T 31015-1 31014-2 31015-2 31020. 31014-1
qPCR master mixes, Fast Plus EvaGreen®
Nukleinsäure-Reagenzien qPCR/RT-qPCR Enzyme und Kits
A qPCR master mix containing Evagreen® qPCR dye and Cheetah™ Taq hotstart DNA polymerase, suitable for qPCR using a fast cycling protocol.
  • 2X qPCR master mix for fast cycling protocols
  • EvaGreen® dye-based
  • Contains Cheetah™ Taq hotstart DNA polymerase
Fast Plus EvaGreen® qPCR Master Mix contains Evagreen® qPCR dye and Cheetah™ Taq hotstart DNA polymerase. It is suitable for qPCR using a fast cycling protocol. However for most PCR applications we recommend the Forget-Me-Not™ series of master mixes, for superior activity.
Fast Plus Evagreen® qPCR Master Mix is a 2X master mix, and is supplied with a separate vial of ROX reference dye, for use with high ROX, low ROX, or no ROX instruments.