Storage drums
Lieferant: Sampling Systems
SAMPA126-25EA 1080 EUR
SAMPA126-25 300-0622 300-0623 300-0624 300-0625 300-0626 SAMPA126-05 SAMPA126-15 SAMPA126-50 SAMPA126-10 SAMPA126-20 SAMPA126-30 300-0620 300-0621
Storage drums
Fässer, Fässer, Trommeln
These drums are used for the storage and processing of powders and liquids.
  • Open top pharmaceutical grade drums with lids
  • Made of 316L stainless steel body and lid
  • Supplied with side handles
The robust, hygienic design means they are suitable for the most demanding applications. These drums are finished to a bright polish finish with all the welds ground and polished.
Lieferumfang: Supplied with lid, clamp and O-ring.