Natriumhydroxid 97,0-100,5%, Plätzchen Ph. Eur., NF, BP, JP

Lieferant: VWR Chemicals
Gefahr

28248.367EA 221 EUR
28248.367 28248.460 28248.298
Natriumhydroxid 97,0-100,5%, Plätzchen Ph. Eur., NF, BP, JP
Sodium hydroxide

Appearance: White or almost white, crystalline masses, supplied as pellets, sticks or slabs, deliquescent, readily absorbing carbon dioxide.


Bitte beachten Not for use as Active Pharmaceutical Ingredient (API)

Formel: NaOH
Molecular Weight: 40 g/mol
Siedepunkt: 1390 °C (1013 hPa)
Schmelzpunkt: 323 °C
Dichte: 2,13 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003548
CAS-Nummer: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8,II
REACH: 01-2119457892-27
Merck Index: 14,08627

Bestellen

Spezifikation Testergebnisse

NF specifications:
Assay (total alkali) [NF] 95.0 --> 100.5 %
Content of sodium [NF] 54.0 --> 59.8 %
Identification B Passes test NF
Insoluble substances and organic matter Passes test NF
JP specifications:
Assay (calculated as NaOH) [JP] Min. 95.0 %
Identification 1 Passes test JP
Identification 2 Passes test JP
Clarity and colour of solution Passes test JP
Cl (Chloride) [JP] Max. 0.050 %
Heavy metals (as Pb) [JP] Max. 30 ppm
K (Potassium) Passes test JP
Sodium carbonate [JP] Max. 2.0 %
Hg (Mercury) Passes test JP

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR