Clamps for conical standard ground joints, KECK™, DURAN®
Lieferant: DWK Life Sciences
KECK™
271-7107EA 4.83 EUR
271-7107 271-7108 271-7116 271-7117 271-7111 271-7114 271-7115 271-7112 271-7113
Clamps for conical standard ground joints, KECK™, DURAN®
Klemmen Schliffklemmen
POM. These clips made of temperature resistant polyacetal (POM) are easy, safe and can be fitted quickly.
  • Good spring properties
  • Colour coded by size
  • No risk of scratching the glass
  • Will not corrode or bend out of shape