n-Hexan ≥99%, AnalaR NORMAPUR® analytisches Reagens

Lieferant: VWR Chemicals
Gefahr

24608.460EA 1090 EUR
24608.460 24608.296 24608.321
n-Hexan ≥99%, AnalaR NORMAPUR® analytisches Reagens
n-Hexan
Formel: H₃C(CH₂)₄CH₃
Molecular Weight: 86,18 g/mol
Siedepunkt: 69 °C (1013 hPa)
Schmelzpunkt: –94,3 °C
Dichte: 0,659 g/cm³ (20 °C)
Flammpunkt: –22 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00009520
CAS-Nummer: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3,II
REACH: 01-2119480412-44
Merck Index: 13,04712
https://de.vwr.com/stibo/web/std.lang.all/70/18/737018.jpg

Bestellen

Spezifikation Testergebnisse

Assay Min. 99.0 %
Acidity Max. 0.0004 meq/g
Colouration Max. 10 APHA
Density (20/4) 0.658 - 0.662
Benzene Max. 100 ppm
Evaporation residue Max. 10 ppm
Thiophene Max. 1 ppm
Total S (as SO4) Max. 50 ppm
Water Max. 100 ppm
Al (Aluminium) Max. 0.5 ppm
B (Boron) Max. 0.02 ppm
Ba (Barium) Max. 0.1 ppm
Ca (Calcium) Max. 0.5 ppm
Cd (Cadmium) Max. 0.05 ppm
Co (Cobalt) Max. 0.02 ppm
Cr (Chromium) Max. 0.02 ppm
Cu (Copper) Max. 0.02 ppm
Fe (Iron) Max. 0.1 ppm
Mg (Magnesium) Max. 0.1 ppm
Mn (Manganese) Max. 0.02 ppm
Ni (Nickel) Max. 0.02 ppm
Pb (Lead) Max. 0.1 ppm
Sn (Tin) Max. 0.1 ppm
Zn (Zinc) Max. 0.1 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR