Petroleumbenzin, 60…80 °C, AnalaR NORMAPUR® analytisches Reagens

Lieferant: VWR Chemicals
Gefahr

Synonyme: BenzinPetrolether

23840.326EA 60.6 EUR
23840.326 23840.292 23840.460
Petroleumbenzin, 60…80 °C, AnalaR NORMAPUR® analytisches Reagens
Petroleumbenzin
Siedepunkt: 60…80 °C (1013 hPa)
Dichte: 0,67 g/cm³ (20 °C)
Flammpunkt: –40 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00081849
CAS-Nummer: 64742-49-0
EINECS: 265-151-9
UN: 1268
ADR: 3,II

Bestellen

Spezifikation Testergebnisse

Acidity Max. 0.0002 meq/g
Bromine value Max. 3
Colouration Max. 10 APHA
Substances coloured by H2SO4 Max. 150 APHA
Boiling range (60-80°C) Min. 90 %
Evaporation residue Max. 10 ppm
Sulphur compounds (as CS2) Max. 3 ppm
Water Max. 100 ppm
Al (Aluminium) Max. 0.15 ppm
Ba (Barium) Max. 0.08 ppm
Ca (Calcium) Max. 0.5 ppm
Cd (Cadmium) Max. 0.05 ppm
Co (Cobalt) Max. 0.02 ppm
Cu (Copper) Max. 0.02 ppm
Fe (Iron) Max. 0.1 ppm
K (Potassium) Max. 0.3 ppm
Mg (Magnesium) Max. 0.08 ppm
Mn (Manganese) Max. 0.02 ppm
Na (Sodium) Max. 0.75 ppm
Ni (Nickel) Max. 0.02 ppm
Pb (Lead) Max. 0.08 ppm
Sn (Tin) Max. 0.1 ppm
Sr (Strontium) Max. 0.03 ppm
Zn (Zinc) Max. 0.1 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR