Petroleumbenzin, 40…60 °C, AnalaR NORMAPUR® ACS analytisches Reagens (max. 0,01% aromatische Kohlenwasserstoffe)

Lieferant: VWR Chemicals
Gefahr

Synonyme: BenzinPetrolether

VWRC23835.363_DEA 0 EUR
VWRC23835.363_D 23835.363 23835.294 23835.460 23835.328 23835.556
Petroleumbenzin, 40…60 °C, AnalaR NORMAPUR® ACS analytisches Reagens (max. 0,01% aromatische Kohlenwasserstoffe)
Petroleumbenzin
Siedepunkt: 40…60 °C (1013 hPa)
Dichte: 0,645…0.665 g/cm³ (20 °C)
Flammpunkt: –40 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00081849
CAS-Nummer: 64742-49-0
EINECS: 265-151-9
UN: 1268
ADR: 3,II
REACH: 01-2119474679-18

Bestellen

Spezifikation Testergebnisse

Acidity Max. 0.0002 meq/g
Bromine value Max. 1
Colouration Max. 10 APHA
Boiling range (40-60°C) (Vol %) Min. 90%
Substances coloured by H2SO4 Max. 150 APHA
Benzene Max. 100 ppm
Evaporation residue Max. 10 ppm
Total S (as SO4) Max. 50 ppm
Water Max. 100 ppm
Al (Aluminium) Max. 0.2 ppm
Ba (Barium) Max. 0.1 ppm
Ca (Calcium) Max. 0.5 ppm
Cd (Cadmium) Max. 0.05 ppm
Co (Cobalt) Max. 0.02 ppm
Cr (Chromium) Max. 0.02 ppm
Cu (Copper) Max. 0.02 ppm
Fe (Iron) Max. 0.1 ppm
K (Potassium) Max. 0.3 ppm
Mg (Magnesium) Max. 0.1 ppm
Mn (Manganese) Max. 0.02 ppm
Na (Sodium) Max. 1 ppm
Ni (Nickel) Max. 0.02 ppm
Pb (Lead) Max. 0.1 ppm
Sn (Tin) Max. 0.1 ppm
Sr (Strontium) Max. 0.03 ppm
Zn (Zinc) Max. 0.1 ppm
Conforms to ACS Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR