Dichlormethan ≥99,5% stabilisiert, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytisches Reagens

Lieferant: VWR Chemicals
Achtung

Synonyme: MethylenchloridChlormethylen

Stabilisierung: Stabilisiert mit 2-Methyl-2-buten (iso-Amylen) 20 ppm

23366.555EA 0 EUR
23366.555 23366.327 23366.460 23366.293
Dichlormethan ≥99,5% stabilisiert, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytisches Reagens
Dichlormethan
Formel: CH₂Cl₂
Molecular Weight: 84,93 g/mol
Siedepunkt: 39,8 °C (1013 hPa)
Schmelzpunkt: –95 °C
Dichte: 1,33 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00000881
CAS-Nummer: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1,III
REACH: 01-2119480404-41
Merck Index: 12,06140
https://de.vwr.com/stibo/web/std.lang.all/14/30/20961430.jpg

Bestellen

Spezifikation Testergebnisse

Assay (on anhydrous substance) Min. 99.5 %
Conforms to ACS Passes test
Conforms to Reag. Ph.Eur. Passes test
IR Spectrum Passes test
Acidity Max. 0.0003 meq/g
Boiling point 39 - 40.5 °C
Colouration Max. 10 APHA
Density (20/4) 1.322 - 1.327
Substances coloured by H2SO4 Max. 60 APHA
Carbon tetrachloride Max. 100 ppm
Chloroform Max. 100 ppm
Evaporation residue Max. 10 ppm
Free halogens Max. 0.3 ppm
2-Methyl-2-butene 10 - 55 ppm
Water Max. 0.02 %
Al (Aluminium) Max. 0.5 ppm
B (Boron) Max. 0.02 ppm
Ba (Barium) Max. 0.1 ppm
Ca (Calcium) Max. 0.5 ppm
Cd (Cadmium) Max. 0.05 ppm
Co (Cobalt) Max. 0.02 ppm
Cr (Chromium) Max. 0.02 ppm
Cu (Copper) Max. 0.02 ppm
Fe (Iron) Max. 0.1 ppm
Mg (Magnesium) Max. 0.1 ppm
Mn (Manganese) Max. 0.02 ppm
Ni (Nickel) Max. 0.02 ppm
Pb (Lead) Max. 0.1 ppm
Sn (Tin) Max. 0.1 ppm
Zn (Zinc) Max. 0.1 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR