Dichlormethan ≥99,5% stabilisiert, AnalaR NORMAPUR® analytisches Reagens
Lieferant: VWR Chemicals
|
Achtung
|
Synonyme:
Methylenchlorid
, Chlormethylen
Stabilisierung:
Stabilisiert mit Ethanol 0,2 %
23354.326EA
66
EUR
23354.326
23354.292
23354.550
23354.460
23354.950
Dichlormethan ≥99,5% stabilisiert, AnalaR NORMAPUR® analytisches Reagens
Dichlormethan
Formel:
CH₂Cl₂ Molecular Weight: 84,93 g/mol Siedepunkt: 39,8 °C (1013 hPa) Schmelzpunkt: –95 °C Dichte: 1,322 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000881 CAS-Nummer: 75-09-2 EINECS: 200-838-9 UN: 1593 ADR: 6.1,III REACH: 01-2119480404-41 Merck Index: 12,06140 |
Spezifikation Testergebnisse
Assay (on anhydrous substance) | Min. 99.5 % |
Acidity | Max. 0.0005 meq/g |
Boiling point | 39.0 - 40.5 °C |
Colouration | Max. 10 APHA |
Density (20/4) | 1.322 - 1.327 |
Substances coloured by H2SO4 | Max. 60 APHA |
Carbon tetrachloride | Max. 100 ppm |
Chloroform | Max. 100 ppm |
Ethanol | 0.15 - 0.25 % |
Evaporation residue | Max. 10 ppm |
Free chlorine | Max. 0.3 ppm |
Water | Max. 0.02 % |
Al (Aluminium) | Max. 0.05 ppm |
B (Boron) | Max. 0.01 ppm |
Ba (Barium) | Max. 0.01 ppm |
Ca (Calcium) | Max. 0.2 ppm |
Cd (Cadmium) | Max. 0.02 ppm |
Co (Cobalt) | Max. 0.01 ppm |
Cr (Chromium) | Max. 0.02 ppm |
Cu (Copper) | Max. 0.02 ppm |
Fe (Iron) | Max. 0.1 ppm |
Mg (Magnesium) | Max. 0.05 ppm |
Mn (Manganese) | Max. 0.01 ppm |
Ni (Nickel) | Max. 0.02 ppm |
Pb (Lead) | Max. 0.05 ppm |
Sn (Tin) | Max. 0.05 ppm |
Zn (Zinc) | Max. 1 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...