Barrels with lid and clamping ring
Lieferant: Rixius
216-5378EA 84.9 EUR
216-5378 216-1628 216-1627 216-5660 RIXI5-2103-150-MA
Barrels with lid and clamping ring
Fässer, Fässer, Trommeln
HDPE, blue.
  • 30 and 60 litre versions include handles
  • Good chemical and temperature resistance
  • All sizes with UN approval for solids
Heavy duty open top drums with PE lid and galvanised clamping ring. Suitable for storage, disposal and transport of laboratory materials.