These straight sided wide-mouth HDPE jar allows for easy filling and sample retreival. Recommended for collecting soil, sediment and sludge samples to be analyzed for inorganic trace analytes.
- With white LDPE-lined PP cap
- Caps assembled onto containers
- Individually barcoded with lot number and unique container number
Standards: Certified to meet or exceed EPA perfomance-based standards for metals, cyanide and fluoride.
Lieferumfang: Supplied with Certificate of Analysis (CofA) in the case and barcoded with lot number for traceability.
Verpackung: Carton sealed with security tape providing tamper evidence.
Bitte beachten *Produkte auf dieser Seite sind außerhalb des Anwendungsbereichs der EU Richtlinie 2019/904