Silbernitrat ≥99,5%, GPR RECTAPUR®
Lieferant: VWR Chemicals
|
Gefahr
|
Synonyme:
Höllenstein
21570.136EA
160
EUR
21570.136
21570.295
21570.182
21570.238
Silbernitrat ≥99,5%, GPR RECTAPUR®
Silbernitrat
Formel:
AgNO₃ Molecular Weight: 169,87 g/mol Siedepunkt: 444 °C (1013 hPa) Schmelzpunkt: 210 °C Dichte: 4,35 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003414 CAS-Nummer: 7761-88-8 EINECS: 231-853-9 UN: 1493 ADR: 5.1,II REACH: 01-2119513705-43 Merck Index: 12,08661 |
Spezifikation Testergebnisse
Assay | Min. 99.5 % |
Foreign heavy metals (as Pb) | Max. 20 ppm |
Not precipitated by HCl (as SO4) | Max. 100 ppm |
Cl (Chloride) | Max. 10 ppm |
SO4 (Sulphate) | Max. 100 ppm |
Fe (Iron) | Max. 10 ppm |
Pb (Lead) | Max. 20 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...