Jars, wide mouth, with thread, WHEATON®
Lieferant: Wheaton
WHEAW216919EA 242 EUR
WHEAW216919 WHEAW216921 WHEAW216920 215-4449 215-4448 215-4457 WHEAW216923 215-4456 WHEAW216922 215-4455 215-4454 215-4453 215-4452 215-4451 215-4450
Jars, wide mouth, with thread, WHEATON®
Dosen Dosen, Universal
Jars, made of clear soda-lime glass, straight sided.
  • Wide mouth design is great for large solid samples
  • Walls allow for complete removal of contents
  • Ideal for soil sampling and environmental applications
  • Available with caps attached or bulk packs without caps
Standards: Conforms to USP Type III requirements.