Dropping bottles, LDPE, Wheaton®
Lieferant: DWK Life Sciences
WHEAW242823-AEA 1790 EUR
WHEAW242823-A WHEAW242822-A WHEAW242821-A WHEAW242826-A WHEAW242825-A WHEAW242824-A 215-4149 215-4148 215-4147 215-4146 215-4145 215-4144 215-4143 215-4142 215-4141 215-4140 WHEAW242834-A WHEAW242833-A WHEAW242832-A WHEAW242831-A WHEAW242836-A WHEAW242835-A 215-4139 215-4138 215-4137 215-4136
Dropping bottles, LDPE, Wheaton®
Flaschen Tropfflaschen
Dropping bottles made of natural or white LDPE. The white LDPE bottles tends to protect the sensitive samples from UV light.
  • Good chemical resistance
  • Good temperature resistance