Messzylinder, DURAN®, Klasse B
Lieferant: DWK Life Sciences
215-3647EA 99.64 EUR
215-3647 215-3646 215-3645 215-3644 215-3643 215-3642 215-3641 215-3640 215-3639
Messzylinder, DURAN®, Klasse B
Zylinder Messzylinder
DURAN®, Borosilikatglas 3.3, klar., Klasse B.
  • Calibrate to contain ('In')
  • Marks and inscription in white
  • With spout
  • With large hexagonal base
Base ist equipped with three knobs to increase the stability
Standards: DIN ISO 4788