Messzylinder, kurze Form, DURAN®
Lieferant: DWK Life Sciences
215-3638EA 105.01 EUR
215-3638 215-3637 215-3636 215-3635 215-3634 215-3633 215-3632 215-3631
Messzylinder, kurze Form, DURAN®
Zylinder Messzylinder
DURAN®, borosilicate glass 3.3, clear, class B, low form.
  • Calibrate to contain ('In')
  • Marks and inscription in white
  • With spout
  • With large hexagonal base
Base ist equipped with three knobs to increase the stability