Spray bottles
Lieferant: Semadeni
215-0215EA 12.9 EUR
215-0215 SEMA1055 215-0281
Spray bottles
Flaschen Sprühflaschen
Robust and handy LDPE spray bottles with stable construction.
  • Emitted volume by pressure 1,3 ml
  • Neck Ø 25 mm
  • Good chemical resistance even against aggressive media
The spray head and nozzle are made of PP and PE. By rotation, the nozzle can be closed or adjusted to produce a very fine vapour through to a powerful jet.