Dosing bottles, LDPE
Lieferant: Semadeni
SEMA1210EA 8.4 EUR
SEMA1210 215-0291 215-0280
Dosing bottles, LDPE
Flaschen Tropfflaschen
LDPE, translucent dosing bottles attached white PP cap with pouring spout.
  • Good chemical resistance
  • Good temperature resistance
Ideal for fast and clean dosing and sampling of the desired liquid quantity.
Bitte beachten Not suitable for shipping liquids.