Trays, multipurpose, PP
Lieferant: Semadeni
SEMA3618EA 72.2 EUR
SEMA3618 SEMA3619 215-0272 SEMA3614 SEMA3615 SEMA3617
Trays, multipurpose, PP
Schalen/Schüsseln/Töpfe Laborschalen und Laborwannen
Made of PP, white.
  • Stable, durable, unbreakable
  • Temperature resistant from −20 to +130 °C
  • Smooth surfaces for easy cleaning
Multipurpose trays. Suitable for use as a tray for aggressive substances, sterilisation and photographic development.