Laboratory trays, HDPE
Lieferant: Semadeni
216-7348EA 24.7 EUR
216-7348 215-0235
Laboratory trays, HDPE
Schalen/Schüsseln/Töpfe Laborschalen und Laborwannen
Laboratory trays made of white HDPE.
  • Stackable
  • Good chemical resistance
  • Good temperature resistance
With reinforcing ribs and supporting edge.