Drosophila containers
Lieferant: Greiner bio-one
212-3171EA 194 EUR
212-3171 525-0196
Drosophila containers
Flaschen Drosophila-Flaschen
PS, crystal-clear.
  • For single use only
  • Temperature resistance –20 to +60 °C
  • Without Ceaprene stopper (must be ordered separately)
Ideally suited for the cultivation of Drosophila melanogaster.
  • For single use only
  • Temperature resistance -20 to +60 °C
  • Without Ceaprene stopper (must be ordered separately)
Ideally suited for the cultivation of Drosophila melanogaster.