Salpetersäure 52,5%, AnalaR NORMAPUR® analytisches Reagens

Lieferant: VWR Chemicals
Gefahr

20420.462EA 404 EUR
20420.462 20420.291 20420.325
Salpetersäure 52,5%, AnalaR NORMAPUR® analytisches Reagens
Salpetersäure
Formel: HNO₃
Molecular Weight: 63,01 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00011349
CAS-Nummer: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8,II

Bestellen

Spezifikation Testergebnisse

Assay 52.5 - 54.0 %
Density (20/4) 1.325 - 1.335
Heavy metals (as Pb) Max. 2 ppm
Ignition residue (SO4) Max. 10 ppm
Cl (Chloride) Max. 0.5 ppm
PO4 (Phosphate) Max. 2 ppm
SO4 (Sulphate) Max. 2 ppm
As (Arsenic) Max. 0.05 ppm
Fe (Iron) Max. 1 ppm
Mn (Manganese) Max. 0.4 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR