Acetonitril ≥99,5%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytisches Reagens
Lieferant: VWR Chemicals
|
Gefahr
|
Synonyme:
Essigsäurenitril
, Ethansäurenitril
, Ethannitril
, Methylcyanid
20071.328EA
104
EUR
20071.328
20071.950
20071.294
20071.550
20071.460
Acetonitril ≥99,5%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytisches Reagens
Acetonitril
Formel:
H₃CCN Molecular Weight: 41,05 g/mol Siedepunkt: 81,6 °C (1013 hPa) Schmelzpunkt: –45,7 °C Dichte: 0,782 g/cm³ (20 °C) Flammpunkt: 5 °C (geschlossener Tiegel) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001878 CAS-Nummer: 75-05-8 EINECS: 200-835-2 UN: 1648 ADR: 3,II REACH: 01-2119471307-38 Merck Index: 13,00071 |
Spezifikation Testergebnisse
Assay (on anhydrous substance) | Min. 99.5 % |
IR Spectrum | Passes test |
Solution (100 g/l) | Passes test |
Acidity | Max. 0.0002 meq/g |
Alkalinity | Max. 0.0006 meq/g |
Boiling point | 80 - 82 °C |
Colouration | Max. 10 APHA |
Density (20/4) | 0.781 - 0.784 |
Density (20/20) | 0.782 - 0.785 |
Brechungsindex (20°C) | 1.343 - 1.345 |
Evaporation residue | Max. 10 ppm |
Water | Max. 0.15 % |
CN (Cyanide) | Max. 50 ppm |
Cd (Cadmium) | Max. 0.02 ppm |
Co (Cobalt) | Max. 0.01 ppm |
Cu (Copper) | Max. 0.04 ppm |
Fe (Iron) | Max. 0.05 ppm |
Mn (Manganese) | Max. 0.01 ppm |
Ni (Nickel) | Max. 0.02 ppm |
Pb (Lead) | Max. 0.05 ppm |
Zn (Zinc) | Max. 0.1 ppm |
Transmittance (255-420 nm) | Min. 98 % |
Conforms to ACS | Passes test |
Conforms to Reag. Ph.Eur. | Passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...