Ammoniaklösung 10%, AnalaR NORMAPUR®

Lieferant: VWR Chemicals
Gefahr

Synonyme: AmmoniakAmmoniumhydroxid

190063UEA 135 EUR
190063U 190064V VWRC190065W
Ammoniaklösung 10%, AnalaR NORMAPUR®
Ammoniaklösung
Formel: NH₃
Molecular Weight: 35,05 g/mol
Dichte: 0,957 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00066650
CAS-Nummer: 1336-21-6
EINECS: 215-647-6
UN: 2672
ADR: 8,III

Bestellen

Spezifikation Testergebnisse

Gehalt 9,0 - 11,0 %
Appearance Clear colourless liquid
Cl (Chloride) Max. 0.5 ppm
PO₄ (Phosphate) Max. 0.1 ppm
S (Sulphide) Max. 0.1 ppm
SO₄ (Sulphate) Max. 0.1 ppm
Fe (Iron) Max. 0.1 ppm
K (Potassium) Max. 1 ppm
Mg (Magnesium) Max. 0.5 ppm
Mn (Manganese) Max. 0.1 ppm
Na (Sodium) Max. 1 ppm
Ni (Nickel) Max. 0.05 ppm
Pb (Lead) Max. 0.05 ppm
Zn (Zinc) Max. 0.2 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR