Melting point standards, p-Toluamide, traceable to primary standards, Supelco®
Lieferant: Merck
15809-5GEA 50.1 EUR
15809-5G
Melting point standards, p-Toluamide, traceable to primary standards, Supelco®
Standards Schmelzpunkt-Standards
Mettler-Toledo Calibration substance ME 30130597, p-Toluamide, traceable to primary standards (LGC).
Suitable for dropping point calibration.