ESD-Sicherheitsschuhe, zum Schnüren, uvex 2, vibram®, 6522
Lieferant: Uvex
uvex 2
114-8664EA 125 EUR
114-8664 114-8660 114-8638 114-8656 114-8652 114-8648 114-8064 114-8065 114-8654 114-8072 114-8076 114-8643 114-8666 114-8074 114-8070 114-8060 114-8663 114-8657 114-8659 114-8667 114-8066 114-8063 114-8636 114-8650 114-8073 114-8670 114-8639 114-8061 114-8649 114-8635 114-8668 114-8640 114-8062 114-8067 114-8651 114-8665 114-8662 114-8068 114-8669 114-8655 114-8069 114-8637 114-8658 114-8653 114-8647 114-8642 114-8071 114-8059 114-8646 114-8645 114-8075 114-8641 114-8644 114-8661
ESD-Sicherheitsschuhe, zum Schnüren, uvex 2, vibram®, 6522
Schuhe und Stiefel
Metal-free, lightweight, flexible and robust safety shoe made from breathable, water repellent high-tech microvelour upper, soft padded tongue and collar, and a dual-density slip-resistant VIBRAM® rubber sole resistant to chemicals, oils and gasoline.
  • Climazone technology for improved comfort
  • Breathable distance mesh lining
  • Removable anti-static comfort insole with moisture control system; orthopaedic insoles possible
  • Virtually seam-free construction to eliminate pressure points
  • Lightweight two-layer sole with excellent shock absorption outstanding slip resistance
  • 100% metal-free xenova® toecap, non magnetic (200 J)
  • Injection-moulded PU scuffcap for additional protection
  • Flexible, metal-free puncture resistant midsole
  • Fulfils ESD requirements: Resistance to ground inferior to 35 megaohms
For medium industrial applications including the chemical, transport, logistics, energy and mechanical engineering industries.
Colour: Black/orange
Standards: EN 61340 (ESD); DGUV112-191; EN ISO 20345:2011; S3, HI, HRO, SRC