BioClean RABS and isolator sleeves are manufactured from nitrile with incredibly low levels of particles and excellent ESD properties.
- Nitrile
- Ultra-clean surface ensures product protection
- 100% inspected and air leak tested (prior to being guillotined)
- Sterilised via gamma irradiation
- 90 mm cuff length
Designed for use in product contact areas, BioClean RABS/Isolator sleeves are fully validated for sterility with an SAL (Sterility Assurance Level) of 10⁻⁶ and have a cuff ring diameter of 90 mm to fit most available cuff ring systems.
Suitable for use in ISO Class 4 and EU GMP Grade A environments. Three years from the date of manufacture. Store in a dry, cool place away from sunlight and florescent light.
Standards: ISO9001:2015 and PPER 2016/425, ASTM D6978-05, (EC) 1935/2004 and (EC) 2023/2006 and ISO 11137-1:2006.
Verpackung: Triple bagged: One piece per sealed inner PE bag; one inner bag per sealed second inner PE bag; one second inner bag per sealed outer PE bag; 20 outer bags per lined inner white Correx polyethylene box (20 pieces).
Bitte beachten No glove provides complete protection against all chemicals. Users must test the gloves against the particular chemicals and environment in which they will be used.