Partikelfilternde Einmal-Halbmasken, FFP2/FFP3, Serie 3000 AIR
Lieferant: Moldex
Serie 3000 AIR
111-0592EA 55 EUR
111-0592 111-0542 111-0593 111-1581 111-1582
Partikelfilternde Einmal-Halbmasken, FFP2/FFP3, Serie 3000 AIR
Atemschutzmasken Einweg-Atemschutzmasken Atemschutzmasken gegen Partikel
These respiratory masks are available in two sizes, with or without valve, as disposable or multi-use version.
  • Folded Filter Technology with 260% more filter surface area
  • Lower breathing resistance
  • Reduced total mask consumption
  • ActivForm®: Automatically fits to facial contours
  • DuraMesh® outer shell: Durable and longer lasting
  • Easy to put on and off due to clip and loop strap design
Standards: EN 149