Chloride CRM, standard solutions, Supelco®
Lieferant: Merck
1.32231.0100EA 50.8 EUR
1.32231.0100 1.32230.0100 1.33011.0100 1.32229.0100
Chloride CRM, standard solutions, Supelco®
Standards Anionische Standards Chlorid-Standards
NaCl in H₂O
  • Diluted standard solutions that are ready-to-use for accurate measurements
  • Traceable to SRM from NIST and USP
  • Full certificates of analysis with comprehensive quality documentation
  • Information on concentration and expanded measurement uncertainty
  • Save time and minimize errors
Method used for Uncertainty Measurement (Accuracy in the table below)= Photometry
Standards: Traceable to SRM from NIST and USP.