Supelco® COD CRM, standard solutions
Lieferant: Merck
Supelco®
1.25034.0100EA 44.2 EUR
1.25034.0100 1.25028.0100 1.25032.0100 1.25029.0100 1.25033.0100 1.25030.0100 1.25035.0100 1.25031.0100
Supelco® COD CRM, standard solutions
Standards Umweltanalyse-Standards
  • Diluted standard solutions that are ready-to-use for accurate measurements
  • Traceable to SRM from NIST and USP
  • Full certificates of analysis with comprehensive quality documentation
  • Information on concentration and expanded measurement uncertainty
  • Save time and minimize errors
Method used for Uncertainty Measurement (Accuracy in the table below)= Photometry
Standards: Traceable to SRM from NIST and USP.