Ethanol ≥99,9% (durch GC), LiChrosolv®, Gradienten-Qualität für die Flüssigchromatographie, Supelco®
Lieferant: Merck
|
Gefahr
|
M-Clarity™ Qualitätsstufe = MQ100
1.11727.2500EA
228
EUR
1.11727.2500
1.11727.2500DP
1.11727.0500
MERC1.11727.1000DP
1.11727.9185
1.11727.9030
1.11727.4000DP
1.11727.9910
MERC1.11727.4000_T
MERC1.11727.9030_T
1.11727.1000
MERC1.11727.4004_T
MERC1.11727.2500_T
MERC1.11727.1000_T
1.11727.4000
Ethanol ≥99,9% (durch GC), LiChrosolv®, Gradienten-Qualität für die Flüssigchromatographie, Supelco®
Ethanol
With their high degree of UV transmittance, low particle count, low acidity and alkalinity and low evaporation residue level, LiChrosolv® solvents are ideal for reproducible separations. Since separations are normally carried out under gradient conditions in analytical HPLC, we offer solvents in 'gradient grade' as well as 'isocratic grade'. This minimises the gradient effect of the solvent involved.
Formel:
C₂H₅OH Molecular Weight: 46,07 g/mol Siedepunkt: 78,3 °C (1013 hPa) Schmelzpunkt: –117 °C Dichte: 0,7895 g/cm³ (20 °C) Flammpunkt: 12 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003568 CAS-Nummer: 64-17-5 EINECS: 200-578-6 UN: 1170 ADR: 3, REACH: 01-2119457610-43 Merck Index: 12,03806 |
Spezifikation Testergebnisse
Purity (GC) | ≥ 99.9 % |
Identity (IR) | conforms |
Evaporation residue | ≤ 2.0 mg/l |
Water | ≤ 0.1 % |
Acidity | ≤ 0.0002 meq/g |
Alkalinity | ≤ 0.0002 meq/g |
Gradient grade (at 235 nm) | ≤ 5 mAU |
Gradient grade (at 254 nm) | ≤ 2 mAU |
Transmission (at 225 nm) | ≥ 60 % |
Transmission (at 240 nm) | ≥ 85 % |
Transmission (from 260 nm) | ≥ 98 % |
Filtered by 0.2 µm filter |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...