Salzsäure 37%, EMSURE® ACS, ISO, Reag. Ph. Eur. zur Analyse, Supelco®

Lieferant: Merck
Gefahr


M-Clarity™ Qualitätsstufe = MQ300

1.00317.9026EA 292 EUR
1.00317.9026 1.00317.9200 1.00317.1000 1.00317.1011 1.00317.1010 1.00317.2011 1.00317.2501 1.00317.2510 1.00317.0510 1.00317.2500
Salzsäure 37%, EMSURE® ACS, ISO, Reag. Ph. Eur. zur Analyse, Supelco®
Salzsäure
Formel: HCl
Molecular Weight: 36,46 g/mol
Siedepunkt: 110 °C (1013 hPa)
Schmelzpunkt: –30 °C
Dichte: 1,19 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011324
CAS-Nummer: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II

Bestellen

Spezifikation Testergebnisse

Assay (alkalimetric) 37.0 - 38.0 %
Identity passes test
Color ≤ 10 Hazen
Appearance passes test
Appearance of solution passes test
Bromide (Br) ≤ 50 ppm
Free chlorine (Cl) ≤ 0.4 ppm
Phosphate (PO₄) ≤ 0.5 ppm
Sulfate (SO₄) ≤ 0.5 ppm
Sulfite (SO₃) ≤ 0.5 ppm
Heavy metals (as Pb) ≤ 1 ppm
Ag (Silver) ≤ 0.020 ppm
Al (Aluminium) ≤ 0.050 ppm
As (Arsenic) ≤ 0.010 ppm
Au (Gold) ≤ 0.050 ppm
B (Boron) ≤ 0.100 ppm
Ba (Barium) ≤ 0.010 ppm
Be (Beryllium) ≤ 0.010 ppm
Bi (Bismuth) ≤ 0.050 ppm
Ca (Calcium) ≤ 0.300 ppm
Cd (Cadmium) ≤ 0.010 ppm
Co (Cobalt) ≤ 0.010 ppm
Cr (Chromium) ≤ 0.010 ppm
Cu (Copper) ≤ 0.010 ppm
Fe (Iron) ≤ 0.100 ppm
Ga (Gallium) ≤ 0.050 ppm
Ge (Germanium) ≤ 0.020 ppm
Hg (Mercury) ≤ 0.010 ppm
K (Potassium) ≤ 0.100 ppm
Li (Lithium) ≤ 0.010 ppm
Mg (Magnesium) ≤ 0.050 ppm
Mn (Manganese) ≤ 0.010 ppm
Mo (Molybdenum) ≤ 0.010 ppm
NH₄ (Ammonium) ≤ 1 ppm
Na (Sodium) ≤ 0.300 ppm
Ni (Nickel) ≤ 0.020 ppm
Pb (Lead) ≤ 0.010 ppm
Pt (Platinum) ≤ 0.100 ppm
Sn (Tin) ≤ 0.050 ppm
Sr (Strontium) ≤ 0.010 ppm
Ti (Titanium) ≤ 0.020 ppm
Tl (Thallium) ≤ 0.020 ppm
V (Vanadium) ≤ 0.010 ppm
Zn (Zinc) ≤ 0.050 ppm
Zr (Zirconium) ≤ 0.020 ppm
Residue on ignition (as SO₄) ≤ 3 ppm
Non volatile matter ≤ 10 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR