Farbreferenzlösungen, gebrauchsfertig, gemäß Ph Eur., Certipur®, Supelco®
Lieferant: Merck
1.00268.0001EA 450 EUR
1.00268.0001 1.00265.0001 1.00269.0001 1.00266.0001 1.00267.0001
Farbreferenzlösungen, gebrauchsfertig, gemäß Ph Eur., Certipur®, Supelco®
Standards Farbreferenzen und Spektroskopie-Standards
Certipur® colour reference solutions are developed for ‘Method 1’ as described in chapter 2.2.2 of the European Pharmacopoeia.
  • Stable solutions with a minimum shelf life of 3 years
  • Ready to use solutions
  • Certificate of Analysis included