Redox standards, Supelco®
Lieferant: Merck
78855-500MLEA 106 EUR
78855-500ML 04887-500ML 87783-500ML 40661-500ML 13198-500ML 80138-500ML 77088-500ML
Redox standards, Supelco®
Standards Redox-/ORP-Standards
The values are the potential of a platinum electrode versus an Ag/AgCl reference in 3M potassium chloride.