Vanderzant Modification Vitamin Mixture
Lieferant: MP Biomedicals
0290324405.EA 1198 EUR
0290324405. ICNA0290324401
Vanderzant Modification Vitamin Mixture
Tierbedarf Tiernahrung
Vanderzant vitamin mixture for insects is an animal-derived component free vitamin mixture.
  • Mixture of 12 vitamins
  • Powdered
Developed for use in insect cell culture.